COMH4211AB9A Combi-Blocks, Inc.

3-Hydroxy-2,3-dihydro-1h-inden-1-one

  • MDL number MFCD11036278
  • Cas Number : 26976-59-0
  • Molecular Weight : 148.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1340

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1340
Availability details for
Quantity
  • Availability
  • Add to Cart