COMH4211ABA0 Combi-Blocks, Inc.

3-Benzyloxy-2-methyl-benzoic acid

  • MDL number MFCD22041842
  • Cas Number : 208986-50-9
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1347

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1347
Availability details for
Quantity
  • Availability
  • Add to Cart