COMH4211ABA2 Combi-Blocks, Inc.

3-Chloro-2-(2,2,2-trifluoroethoxy)aniline

  • MDL number MFCD11132423
  • Cas Number : 782414-07-7
  • Molecular Weight : 225.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-2-(2,2,2-trifluoroethoxy)aniline
Inchi Code 1S/C8H7ClF3NO/c9-5-2-1-3-6(13)7(5)14-4-8(10,11)12/h1-3H,4,13H2
Inchi Key LLBJPWSONYEMSY-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1349

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1349
Availability details for
Quantity
  • Availability
  • Add to Cart