COMH4211ABA6 Combi-Blocks, Inc.

2-Methyl-4-(propane-2-sulfonyl)-benzoic acid

  • MDL number MFCD30290508
  • Cas Number : 1346608-66-9
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1359

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1359
Availability details for
Quantity
  • Availability
  • Add to Cart