COMH4211ABAA Combi-Blocks, Inc.

2-Amino-4,5-difluoro-n-methylbenzamide

  • MDL number MFCD16159058
  • Cas Number : 1042434-87-6
  • Molecular Weight : 186.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1363

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1363
Availability details for
Quantity
  • Availability
  • Add to Cart