COMH4211ABB2 Combi-Blocks, Inc.

1-[3-(2,2-Difluoro-propoxy)-phenyl]-ethanone

  • MDL number MFCD28530155
  • Cas Number : 1780433-37-5
  • Molecular Weight : 214.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1369

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1369
Availability details for
Quantity
  • Availability
  • Add to Cart