COMH4211ABB4 Combi-Blocks, Inc.

Malonic acid 2,5-dioxo-pyrrolidin-1-yl ester ethyl ester

  • MDL number MFCD22193884
  • Cas Number : 1428131-72-9
  • Molecular Weight : 229.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1371

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1371
Availability details for
Quantity
  • Availability
  • Add to Cart