COMH4211ABB6 Combi-Blocks, Inc.

1-Butyl-5-fluoro-1h-indole-2-carboxylic acid

  • MDL number MFCD23162174
  • Cas Number : 1391467-35-8
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1372

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1372
Availability details for
Quantity
  • Availability
  • Add to Cart