COMH4211ABBA Combi-Blocks, Inc.

1-[3-Methyl-4-(2,2,2-trifluoro-ethoxy)-phenyl]-ethanone

  • MDL number MFCD26442813
  • Cas Number : 1476027-18-5
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1375

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1375
Availability details for
Quantity
  • Availability
  • Add to Cart