COMH4211ABBC Combi-Blocks, Inc.

2-(4-Bromo-2-fluoro-phenyl)-1-morpholin-4-yl-ethanone

  • MDL number MFCD28403614
  • Cas Number : 1092563-30-8
  • Molecular Weight : 302.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1376

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1376
Availability details for
Quantity
  • Availability
  • Add to Cart