COMH4211ABBE Combi-Blocks, Inc.

3-Chloro-4-(2,2-difluoro-propoxy)-benzoic acid

  • MDL number MFCD31539543
  • Cas Number : 1476028-07-5
  • Molecular Weight : 250.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1377

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1377
Availability details for
Quantity
  • Availability
  • Add to Cart