COMH4211ABC0 Combi-Blocks, Inc.

4-Amino-2-ethoxy-n-methylbenzamide

  • MDL number MFCD19640846
  • Cas Number : 1676-82-0
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1378

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1378
Availability details for
Quantity
  • Availability
  • Add to Cart