COMH4211ABC4 Combi-Blocks, Inc.

Methoxy[(2-methylphenyl)methyl]amine

  • MDL number MFCD21100672
  • Cas Number : 543731-13-1
  • Molecular Weight : 151.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1381

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1381
Availability details for
Quantity
  • Availability
  • Add to Cart