COMH4211ABC6 Combi-Blocks, Inc.

Benzyl 4-nitropiperidine-1-carboxylate

  • MDL number MFCD18633013
  • Cas Number : 1268520-00-8
  • Molecular Weight : 264.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1382

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1382
Availability details for
Quantity
  • Availability
  • Add to Cart