COMH4211ABCA Combi-Blocks, Inc.

5-(2,2-Difluoroethoxy)pyridine-2-carboxylic acid

  • MDL number MFCD11846426
  • Cas Number : 1097730-45-4
  • Molecular Weight : 203.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1386

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1386
Availability details for
Quantity
  • Availability
  • Add to Cart