COMH4211ABCC Combi-Blocks, Inc.

(3-Amino-4-fluorophenyl)(morpholino)methanone

  • MDL number MFCD11937887
  • Cas Number : 959795-65-4
  • Molecular Weight : 224.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-5-(4-morpholinylcarbonyl)aniline
Inchi Code 1S/C11H13FN2O2/c12-9-2-1-8(7-10(9)13)11(15)14-3-5-16-6-4-14/h1-2,7H,3-6,13H2
Inchi Key AVMNSVOUWXMAHO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1388

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1388
Availability details for
Quantity
  • Availability
  • Add to Cart