COMH4211ABCE Combi-Blocks, Inc.

3-[3,5-Dimethyl-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyrazol-1-ylmethyl]-benzoic acid methyl ester

  • MDL number MFCD31583210
  • Molecular Weight : 370.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1389

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1389
Availability details for
Quantity
  • Availability
  • Add to Cart