COMH4211ABD0 Combi-Blocks, Inc.

5,8-Dioxa-2,11-diazatetradecanoic acid, 14-(2,5-dihydro-2,5-dioxo-1h-pyrrol-1-yl)-12-oxo-, 1,1-dimethylethyl ester

  • MDL number MFCD31381647
  • Cas Number : 1801876-84-5
  • Molecular Weight : 399.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1390

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1390
Availability details for
Quantity
  • Availability
  • Add to Cart