COMH4211ABD2 Combi-Blocks, Inc.

N-(3-(Piperidin-4-yl)phenyl)propane-2-sulfonamide

  • MDL number MFCD31579318
  • Cas Number : 1799602-69-9
  • Molecular Weight : 282.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1392

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1392
Availability details for
Quantity
  • Availability
  • Add to Cart