COMH4211ABD6 Combi-Blocks, Inc.

4-Isopropoxy-3-methyl-benzylamine

  • MDL number MFCD12805196
  • Cas Number : 1183725-82-7
  • Molecular Weight : 179.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1402

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1402
Availability details for
Quantity
  • Availability
  • Add to Cart