COMH4211ABD7 Combi-Blocks, Inc.

2-(4-Hydroxybutyloxy)acetic acid tert-butyl ester

  • MDL number MFCD31618926
  • Cas Number : 475086-57-8
  • Molecular Weight : 204.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1403

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1403
Availability details for
Quantity
  • Availability
  • Add to Cart