COMH4211ABD9 Combi-Blocks, Inc.

6-(3-Chloro-4-fluorophenoxy)pyridin-3-amine

  • MDL number MFCD18899952
  • Cas Number : 1276556-19-4
  • Molecular Weight : 238.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1407

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1407
Availability details for
Quantity
  • Availability
  • Add to Cart