COMH4211ABDA Combi-Blocks, Inc.

4-Bromo-1-methyl-2-(2,2,2-trifluoro-ethoxy)-benzene

  • MDL number MFCD31557879
  • Cas Number : 1691774-44-3
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1409

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1409
Availability details for
Quantity
  • Availability
  • Add to Cart