COMH4211ABDC Combi-Blocks, Inc.

3-[(2-Methylpropyl)sulfanyl]propan-1-ol

  • MDL number MFCD12180134
  • Cas Number : 1155607-49-0
  • Molecular Weight : 148.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1417

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1417
Availability details for
Quantity
  • Availability
  • Add to Cart