COMH4211ABDE Combi-Blocks, Inc.

2-Tritylsulfanyl-benzoic acid

  • MDL number MFCD09053458
  • Cas Number : 943423-30-1
  • Molecular Weight : 396.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1420

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1420
Availability details for
Quantity
  • Availability
  • Add to Cart