COMH4211ABDF Combi-Blocks, Inc.

4-[2-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenoxymethyl]-benzoic acid methyl ester

  • MDL number MFCD31614102
  • Molecular Weight : 368.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1423

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1423
Availability details for
Quantity
  • Availability
  • Add to Cart