COMH4211ABE0 Combi-Blocks, Inc.

Hydrocinnamic acid n-hydroxysuccinimide ester

  • MDL number MFCD09840689
  • Cas Number : 109318-10-7
  • Molecular Weight : 247.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1430

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1430
Availability details for
Quantity
  • Availability
  • Add to Cart