COMH4211ABE2 Combi-Blocks, Inc.

(3-Benzyloxy-2-methyl-phenyl)-methanol

  • MDL number MFCD22041843
  • Cas Number : 918524-13-7
  • Molecular Weight : 228.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1433

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1433
Availability details for
Quantity
  • Availability
  • Add to Cart