COMH4211ABE3 Combi-Blocks, Inc.

2-(2-Bromoethoxy)naphthalene

  • MDL number MFCD00837561
  • Cas Number : 13247-80-8
  • Molecular Weight : 251.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2-bromoethoxy)naphthalene
Inchi Code 1S/C12H11BrO/c13-7-8-14-12-6-5-10-3-1-2-4-11(10)9-12/h1-6,9H,7-8H2
Inchi Key NKALVESVNGTFHZ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1435

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1435
Availability details for
Quantity
  • Availability
  • Add to Cart