COMH4211ABE6 Combi-Blocks, Inc.

11-Methoxy-11-oxoundecanoic acid

  • MDL number MFCD00673464
  • Cas Number : 3927-60-4
  • Molecular Weight : 230.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 11-methoxy-11-oxoundecanoic acid
Inchi Code 1S/C12H22O4/c1-16-12(15)10-8-6-4-2-3-5-7-9-11(13)14/h2-10H2,1H3,(H,13,14)
Inchi Key JAELIPKEGUQPKD-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1445

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1445
Availability details for
Quantity
  • Availability
  • Add to Cart