COMH4211ABE7 Combi-Blocks, Inc.

N-(5-Bromo-2-nitrophenyl)-2-chloropyrimidin-4-amine

  • MDL number MFCD29044942
  • Cas Number : 1394119-49-3
  • Molecular Weight : 329.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1447

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1447
Availability details for
Quantity
  • Availability
  • Add to Cart