COMH4211ABE8 Combi-Blocks, Inc.

2-(1-Ethanesulfonyl-1h-pyrazol-4-yl)-pyrimidin-4-ylamine

  • MDL number MFCD31583051
  • Cas Number : 1612173-67-7
  • Molecular Weight : 253.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1448

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1448
Availability details for
Quantity
  • Availability
  • Add to Cart