COMH4211ABE9 Combi-Blocks, Inc.

2-Ethyl-5-iodobenzoic acid

  • MDL number MFCD18411966
  • Cas Number : 1261451-95-9
  • Molecular Weight : 276.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1450

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1450
Availability details for
Quantity
  • Availability
  • Add to Cart