COMH4211ABEA Combi-Blocks, Inc.

Ethyl 3-amino-4-(1-piperidinyl)benzoate

  • MDL number MFCD00238972
  • Cas Number : 71254-74-5
  • Molecular Weight : 248.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1454

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1454
Availability details for
Quantity
  • Availability
  • Add to Cart