COMH4211ABEC Combi-Blocks, Inc.

5-Bromo-2-ethoxy-n-ethyl-benzamide

  • MDL number MFCD07551367
  • Cas Number : 882080-55-9
  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1460

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1460
Availability details for
Quantity
  • Availability
  • Add to Cart