COMH4211ABED Combi-Blocks, Inc.

4-(3-Methoxypropyl)-2,2-dimethyl-3-oxo-3,4-dihydro-2h-1,4-benzoxazine-6-carboxylic acid

  • MDL number MFCD31561603
  • Cas Number : 1163280-59-8
  • Molecular Weight : 293.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1462

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1462
Availability details for
Quantity
  • Availability
  • Add to Cart