COMH4211ABEF Combi-Blocks, Inc.

(2R)-1-(Cyclopropanesulfonyl)piperidine-2-carboxylic acid

  • MDL number MFCD21754247
  • Cas Number : 1567879-65-5
  • Molecular Weight : 233.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1465

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1465
Availability details for
Quantity
  • Availability
  • Add to Cart