COMH4211ABF0 Combi-Blocks, Inc.

tert-Butyl 3-(3-aminophenoxy)pyrrolidine-1-carboxylate

  • MDL number MFCD24535417
  • Cas Number : 1193104-14-1
  • Molecular Weight : 278.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1467

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1467
Availability details for
Quantity
  • Availability
  • Add to Cart