COMH4211ABF1 Combi-Blocks, Inc.

5-(tert-Butyl-dimethyl-silanyloxy)-3,3-dimethyl-pentan-1-ol

  • MDL number MFCD31613940
  • Cas Number : 220650-54-4
  • Molecular Weight : 246.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1470

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1470
Availability details for
Quantity
  • Availability
  • Add to Cart