COMH4211ABF4 Combi-Blocks, Inc.

2-Chloro-4-(3-fluoro-benzyl)-5-methyl-pyrimidine

  • MDL number MFCD31583046
  • Cas Number : 2270905-66-1
  • Molecular Weight : 236.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1477

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1477
Availability details for
Quantity
  • Availability
  • Add to Cart