COMH4211ABF5 Combi-Blocks, Inc.

(1-Benzyl-1h-pyrazol-4-yloxy)-acetic acid

  • MDL number MFCD31579327
  • Cas Number : 2270906-24-4
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1478

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1478
Availability details for
Quantity
  • Availability
  • Add to Cart