COMH4211ABF7 Combi-Blocks, Inc.

3-[(Benzothiazol-2-yl)oxymethyl]piperidine

  • MDL number MFCD17058506
  • Cas Number : 876148-06-0
  • Molecular Weight : 248.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1481

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1481
Availability details for
Quantity
  • Availability
  • Add to Cart