COMH4211ABF8 Combi-Blocks, Inc.

2-(3-Chloro-4-fluorophenoxy)-5-nitropyridine

  • MDL number MFCD10224166
  • Cas Number : 1014360-34-9
  • Molecular Weight : 268.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1483

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1483
Availability details for
Quantity
  • Availability
  • Add to Cart