COMH4211ABFA Combi-Blocks, Inc.

[3-Fluoro-5-(propan-2-yloxy)phenyl]methanol

  • MDL number MFCD31628745
  • Cas Number : 1598137-25-7
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1487

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1487
Availability details for
Quantity
  • Availability
  • Add to Cart