COMH4211ABFD Combi-Blocks, Inc.

Ethyl 2-((4-nitrophenoxy)carbonyloxy)acetate

  • MDL number MFCD20483657
  • Cas Number : 236418-39-6
  • Molecular Weight : 269.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1491

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1491
Availability details for
Quantity
  • Availability
  • Add to Cart