COMH4211ABFE Combi-Blocks, Inc.

[2-(3-Chlorophenyl)pyridin-3-yl]methanol

  • MDL number MFCD31561462
  • Cas Number : 450840-05-8
  • Molecular Weight : 219.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1492

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1492
Availability details for
Quantity
  • Availability
  • Add to Cart