COMH4211ABFF Combi-Blocks, Inc.

(2-Chloro-pyridin-3-yl)-cyclopropylmethyl-amine

  • MDL number MFCD12121147
  • Cas Number : 1155017-73-4
  • Molecular Weight : 182.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1493

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1493
Availability details for
Quantity
  • Availability
  • Add to Cart