COMH4211AC01 Combi-Blocks, Inc.

3-(4-Iodo-phenoxy)-azetidine-1-carboxylic acid benzyl ester

  • MDL number MFCD31614016
  • Molecular Weight : 409.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1497

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1497
Availability details for
Quantity
  • Availability
  • Add to Cart