COMH4211AC02 Combi-Blocks, Inc.

1-(6-Aminopyridin-3-yl)azetidin-3-ol

  • MDL number MFCD24627704
  • Cas Number : 1253911-54-4
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1498

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1498
Availability details for
Quantity
  • Availability
  • Add to Cart