COMH4211AC04 Combi-Blocks, Inc.

2-Benzylamino-5-bromo-nicotinic acid tert-butyl ester

  • MDL number MFCD31557858
  • Cas Number : 2208788-33-2
  • Molecular Weight : 363.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1500

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1500
Availability details for
Quantity
  • Availability
  • Add to Cart